Learn More
Macherey-Nagel™ Haloform Test Mixture in n-Pentane
Brand:  Macherey-Nagel™ 722311
Product Code. 10216421
  • 78.35 € / 1mL
Explore more special offers
Specifications
For GC Environmental Analyses
Dichloromethane (795 ng/mL), Trichloromethane (75 ng/mL), 1,1,1-Trichloroethane (67 ng/mL), Tetrachloromethane (80 ng/mL), Trichloroethene (73 ng/mL), Bromodichloromethane (100 ng/mL), Dibromochloromethane (122 ng/mL), Tetrachloroethene (81 ng/mL), Tribromomethane (145 ng/mL)
1 mL
Haloform Test Mixture in n-pentane
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Haloform Test Mixture in n-Pentane > Capacity: 1mL