missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Ham's F-12 Nutrient Mix

Ham's F-12 Nutrient Mix
Brand: Gibco™ 21765029
This item is not returnable.
View return policy
Description
Ham's F-12 Nutrient Mixture (F-12) was designed for serum-free single-cell plating of Chinese Hamster Ovary (CHO) cells. F-12 has since been used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells, including chondrocytes and rat prostate epithelial cells. We offer a variety of Gibco™ F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.Compared to other basal media, F-12 contains a wider variety of components, including zinc, putrescine, hypoxanthine, and thymidine. Serum-free growth of CHO cells in F-12 has led to a variety of improved formulations.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ F-12 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ F-12 product made in our Grand Island, New York facility (11765-054). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Gibco™ F-12 contains no proteins or growth factors. Therefore, F-12 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). F-12 uses a sodium bicarbonate buffer system (1.176 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Specifications
CHO | |
Liquid | |
Glutamine, Phenol Red, Sodium Pyruvate | |
500 mL | |
7.1 to 7.5 | |
Ham's F-12 Nutrient Mix, 500mL, 1 X Concentration, Liquid, 7.1 to 7.5 pH, 12 Months Shelf Life, 280 to 310 mOsm/kg Osmolality, With Sodium Pyruvate, L-glutamine, Phenol Red, 2 to 8°C Storage, For Mammalian Cell Culture |
1 X | |
Ham's F-12 Nutrient Mix | |
Mammalian cell culture | |
ISO 13485 standards, FDA | |
2 to 8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ Ham's F-12 Nutrient Mix > 500mL
Spot an opportunity for improvement?Share a Content Correction