Learn More
Hamilton™ Chaney Adapter Assemb 700 800 7000/1700 1800 Series Microliter/Gastight™ Syring
Designed to work with 700, 800, 7000 series Microliter syringes and 1700 and 1800 series Gastight syringes
Brand:  Hamilton™ 32146
 View more versions of this product
Product Code. 10343213
  • 217.00 € / Each
Explore more special offers
Description
Description
Adapters are designed to give consistent and reproducible injections and also prevent the plungers from bending.
Specifications
Specifications
Reproducibility Adapter
Ea.
Hamilton Syringe Models 800 and 1800 Series