Learn More
Hamilton™ Chaney Adapter Assemb 700 800 7000/1700 1800 Series Microliter/Gastight™ Syring
Designed to work with 700, 800, 7000 series Microliter syringes and 1700 and 1800 series Gastight syringes
103.00 € - 217.00 €
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10281961
|
Hamilton™
14700 |
Hamilton Syringe Models 75 and 701 Series |
166.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13246949
|
Hamilton™
14725 |
Hamilton Syringe Models 702-750, 7000 and 1700 Series |
151.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10015600
|
Hamilton™
14725 |
Hamilton Syringe Models 702-750, 7000 and 1700 Series | N/A | ||||||
10343213
|
Hamilton™
32146 |
Hamilton Syringe Models 800 and 1800 Series |
217.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Adapters are designed to give consistent and reproducible injections and also prevent the plungers from bending.
Specifications
Reproducibility Adapter | |
Ea. |