missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Kleenex™ Everyday Use Hand Cleanser

Dispenses the ideal amount of soap for each wash, preventing overuse. Kleenex™ Everyday Use Hand Cleanser delivers up to twice as many washes per liter as conventional liquid soaps.

Brand:  Kimberly-Clark™ 6331

70.82 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10375582

  • 155.00 € / Pack of 6
Estimated Shipment: 25-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Pink foam soap with a luxury feel and subtle fragrance
  • Supplied in strong, hygienically sealed cassettes that are quick and easy to install
  • Provides up to 1,000 shots of soap,
  • Shrinks to save space when empty and are recyclable (when the pump has been removed).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hand Cleanser
Foam
Dispenser
1000 mL
1.033 kg
Pink
93 x 93 x 180 mm
33.8 oz.
13.6 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kimberly-Clark™ Kleenex™ Everyday Use Hand Cleanser > Color: Pink, Fragrance: Floral, Volume: 1000 mL, Includes: Dispenser

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.