Learn More
Cole-Parmer™ Essentials HandE-Vac Handheld Aspirating System
A complete set of tools for aspiration—rapidly remove supernatant from a variety of cell culture or multi-well plates and dishes with the Argos Technologies HandE-Vac Handheld Aspirator.
226.26 € - 276.00 €
Specifications
Autoclavable Autoclavable
Product Line HandE-Vac
Type Handheld Aspirator
View More Specs
Products 2
Product Code Brand Model Type Price Quantity & Availability  
Product Code Brand Model Type Price Quantity & Availability  
15944991
View Documents
Cole-Parmer™ Essentials
EV514
EV514 Handheld Aspirator
276.00 €
Each
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
15934991
View Documents
Cole-Parmer™ Essentials
EV500
EV500 Handheld Aspirator This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Use with Pasteur pipettes, plastic aspiration pipettes or a variety of adapters for specific applicationsPressure-sensitive button on the top of the unit controls the specific amount of vacuum appliedOperates with standard in-house vacuum source or with the M-Vac or E-Vac aspiration systemsAutoclavable
Specifications
Specifications
Autoclavable
HandE-Vac
Handheld Aspirator