Learn More
Axygen™ Hanging Wafer Robotic Tips: Tecan-Style Liquid Level Sensing Tips, Presterilized
GREENER_CHOICE
Black tips for accurate sample delivery
Brand:  Axygen™ TTF-1000-CBK-HTR-S
 View more versions of this product
Product Code. 12201627
  • 268.67 € / Pack of 1536
Estimated Shipment: 04-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Axygen™ offers a comprehensive line of tips for Tecan™, Aurora Biomed and PerkinElmer™ automation systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Tips are RNase-/DNase-free and nonpyrogenic. Available in nonsterile, sterile, non-filtered and filtered. Clear and Conductive black polypropylene tips are available in 96-well format and is also available in hanging tip rack format designed for Tecan™ Genesis Freedom™/Freedom Eva™/Miniprep with LiHa-1, -4, -8 head, Aurora Biomed Versa™ with 8-Channel head and PerkinElmer™ Janus™/MultiProbe with Variospan-1, 4, 8 head.
  • 1000 μL Liquid Level Sensing Tips
  • 96 Tip format
  • Conductive Black Polypropylene
  • RNase-/DNase-free
  • Nonpyrogenic
  • Sterile
  • Compatibility: Tecan™ Genesis Freedom™/Freedom Eva™/Miniprep with LiHa-1, -4, -8 head, Aurora Biomed Versa™ with 8-Channel head, PerkinElmer™ Janus™/MultiProbe with Variospan-1, 4, 8 head
  • Certifications
    Certified DNAse-, RNAse- and Pyrogen-safe
    TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    1000 μL
    Sterile
    Autoclavable
    For accurate sample delivery
    Automation, Round
    16 Racks/Cs.
    Polypropylene
    Filtered
    Black
    Non-ANSI
    Rack
    Hanging tip rack
    Aurora Biomed Versa 8-channel Head, PerkinElmer MultiProbe Variospan-1, 4, 8 Head