missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Hanging Wafer Robotic Tips: Tecan-Style Liquid Level Sensing Tips, Presterilized
GREENER_CHOICE

Black tips for accurate sample delivery

230.27 € - 447.86 €

Specifications

Sterility Sterile
Color Black
Autoclavable Autoclavable
Certifications/Compliance Non-ANSI
For Use With (Application) For accurate sample delivery
View More Specs

Products 7
Product Code Brand Volume (Metric) Filtered Sterility Tip Style Price Quantity & Availability  
Product Code Brand Volume (Metric) Filtered Sterility Tip Style Price Quantity & Availability  
11394305
GREENER_CHOICE
View Documents
Axygen™
TTF-200-CBK-HTR-S
200 μL Filtered Sterile Automation, Round
434.99 €
Pack of 2304
Estimated Shipment: 04-06-2025
to see stock.
Add to basket
 
11374305
GREENER_CHOICE
View Documents
Axygen™
TTF-20-CBK-HTR-S
20 μL Filtered Sterile Automation, Round
447.86 €
Pack of 2304
Estimated Shipment: 04-06-2025
to see stock.
Add to basket
 
11334305
GREENER_CHOICE
View Documents
Axygen™
TT-50-CBK-HTR-S
50 μL Non-filtered Sterile Automation, Round
378.73 €
Pack of 2304
Estimated Shipment: 04-06-2025
to see stock.
Add to basket
 
11384295
GREENER_CHOICE
View Documents
Axygen™
TT-200-CBK-HTR-S
200 μL Non-filtered Sterile Automation, Round
355.75 €
Pack of 2304
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
12201627
GREENER_CHOICE
View Documents
Axygen™
TTF-1000-CBK-HTR-S
1000 μL Filtered Sterile Automation, Round
268.67 €
Pack of 1536
Estimated Shipment: 04-06-2025
to see stock.
Add to basket
 
11314315
GREENER_CHOICE
View Documents
Axygen™
TTF-50-CBK-HTR-S
50 μL Filtered Sterile Automation, Round
447.86 €
Pack of 2304
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
11384285
GREENER_CHOICE
View Documents
Axygen™
TT-1000-CBK-HTR-S
1000 μL Non-filtered Sterile Automation, Round
230.27 €
Pack of 1536
Estimated Shipment: 04-06-2025
to see stock.
Add to basket
 
Description

Description

Axygen™ offers a comprehensive line of tips for Tecan™, Aurora Biomed and PerkinElmer™ automation systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Tips are RNase-/DNase-free and nonpyrogenic. Available in nonsterile, sterile, non-filtered and filtered. Clear and Conductive black polypropylene tips are available in 96-well format and is also available in hanging tip rack format designed for Tecan™ Genesis Freedom™/Freedom Eva™/Miniprep with LiHa-1, -4, -8 head, Aurora Biomed Versa™ with 8-Channel head and PerkinElmer™ Janus™/MultiProbe with Variospan-1, 4, 8 head.


  • 1000 μL Liquid Level Sensing Tips
  • 96 Tip format
  • Conductive Black Polypropylene
  • RNase-/DNase-free
  • Nonpyrogenic
  • Sterile
  • Compatibility: Tecan™ Genesis Freedom™/Freedom Eva™/Miniprep with LiHa-1, -4, -8 head, Aurora Biomed Versa™ with 8-Channel head, PerkinElmer™ Janus™/MultiProbe with Variospan-1, 4, 8 head
  • Certifications

    Certified DNAse-, RNAse- and Pyrogen-safe

    Specifications

    Specifications

    Sterile
    Autoclavable
    For accurate sample delivery
    Automation, Round
    Black
    Non-ANSI
    Rack
    Polypropylene
    Videos
    SDS
    Documents

    Documents

    Product Certifications
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.