Learn More
Thermo Scientific™ Hastelloy PT1000 External Probe
For Use With: Cimarec and Supernuova Hotplates and Hotplate Stirrers
Brand:  Thermo Scientific™ CIC0001449
77.89 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Product Code. 15493660
  • 97.89 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
For Use With: Cimarec and Supernuova Hotplates and Hotplate Stirrers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hastelloy PT1000 External probe, 0-400°C, Hastelloy, for all sizes of Cimarec+ and SuperNuova+ Hotplates and Hotplate Stirrers
0°C to 400°C
Digital
Cimarec and Supernuova Hotplates and Hotplate Stirrers
Probe
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Hastelloy PT1000 External Probe > PT1000 External probe, 0-400°C