missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Hazard Warning Tape
Used for marking hazardous areas, protruding equipment, or preventing accidents in factories, warehouses and hospitals.
248.00 € - 265.00 €
Specifications
Dimensions (L x W) | 50 mm x 33 m |
---|---|
Length (Metric) | 33 m |
Width (Metric) | 50 mm |
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
17267790
|
3M™
7100015263 |
Black with Yellow Stripes |
265.00 €
24 rolls |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
17277790
|
3M™
7100015266 |
Red |
248.00 €
24 rolls |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
Description
- Use for taping in all non-critical applications
- Conformable backing makes vinyl tape ideal for use on curved and irregular surfaces
- Rubber adhesive provides good instant adhesion on many surfaces
- Maintains vivid colour over time for long-lasting quality
- Can be applied by hand or with floor applicator
- Ideal for marking applications in light traffic areas
Specifications
50 mm x 33 m | |
33 m | |
50 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ Hazard Warning Tape