missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Tank with Spigot

Designed to rationalize laboratory and small production line organization. Kartell™ Tank with Spigot gives the opportunity to optimize water, aqueous solutions and solvents availability in a reduced space.

Brand:  Kartell™ 0177000

 View more versions of this product

Product Code. 10079922

  • 189.00 € / Pack of 5
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made of food grade High Density PE
  • Includes comfortable handle for displacement
  • The spigot is placed in a position allowing the tank to stand, as to facilitate the use of these tanks in packaging and transporting liquids
  • Suitable for foodstuff
Specifications

Specifications

HDPE
380 mm
160 mm
10 L
300 mm
5
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kartell™ Tank with Spigot > Capacity: 10L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.