Learn More
Kartell™ Tank with Spigot
Designed to rationalize laboratory and small production line organization. Kartell™ Tank with Spigot gives the opportunity to optimize water, aqueous solutions and solvents availability in a reduced space.
157.00 € - 189.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10079922
|
Kartell™
0177000 |
10 L |
189.00 €
Pack of 5 |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||||
10640204
|
Kartell™
0177100 |
20 L |
157.00 €
Pack of 4 |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
Description
- Made of food grade High Density PE
- Includes comfortable handle for displacement
- The spigot is placed in a position allowing the tank to stand, as to facilitate the use of these tanks in packaging and transporting liquids
- Suitable for foodstuff
Specifications
HDPE |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Kartell™ Tank with Spigot