missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ HDPE Straining Basket

Perforated straining baskets made from polyethylene plastic allow rapid filling and draining

Brand:  Bel-Art™ SP Scienceware™ H16703-0000

Product Code. 11754133

  • 253.00 € / Each
Estimated Shipment: 25-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Handy for batch rinsing and transporting labware as well as straining or filtering large particles
  • Chemically resistant; withstand acids and alkalis
  • Service temperature up to 82°C (180°F)
  • Perforations are 2mm (0.080 in.)
  • Strap handle rises 10cm (4 in.) above the rim
Specifications

Specifications

Strap Handle
Not Autoclavable
For Dipping Parts or Straining Large Particles
127 mm
2 mm
Straining Basket
Polyethylene
82°C
127 mm
108 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ SP Scienceware™ HDPE Straining Basket > For use with: For Dipping Parts or Straining Large Particles

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.