Learn More
Masterflex™ HDPE T-Connector Barbed Fittings
Barbed T connectors are simple and economical fittings designed for quick and easy assembly of fluid paths in the laboratory.
Brand:  Masterflex™ 30623-67
 View more versions of this product
Product Code. 11745569
  • 25.10 € / Pack of 10
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Use with low-pressure applications, generally less than 30 psi (2 bar)
  • Manufactured from quality HDPE
  • Sterilization by ethylene oxide (ETO) only
  • Complete range of standard sizes available
  • Provided in packs of 10
Specifications
Specifications
Barbed Fitting
3/16 in. tubing I.D.
Ethylene Oxide
HDPE
10 Pack
T-shape