missing translation for 'onlineSavingsMsg'
Learn More

EduLab™ Helical Springs, Slinky

Ideal for studying and demonstrating different types of waves.

Brand:  EduLab™ VTN12302593

 View more versions of this product

Product Code. 13172890

  • 4.74 € / Each
Estimated Shipment: 13-08-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made of metal
  • Available in 50 and 100mm diameter
Specifications

Specifications

75 to 50 mm
Helical
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
EduLab™ Helical Springs, Slinky >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.