missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Economical Hemacytometer with Cover Slips

Economical Hemacytometer with Cover Slips is specially designed for counting erythrocytes and leukocytes.

Brand:  Cole-Parmer™ 79001-00

247.68 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.


Product Code. 11704939

  • 258.00 € / Each
Estimated Shipment: 13-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Two special counting chamber glass coverslips.
  • Light transmission between 29 and 32%.
  • This hemacytometer features high light transmission and brightly illuminated line.
  • Double rulings are visible through a wide range of illumination.
  • Neubauer rulings are 0.1 mm below cover glass.
  • The H-shaped moat offers two counting areas.
  • Fused rhodium on glass provides distinct lines.
  • Two glass coverslips are included.
Specifications

Specifications

Hemacytometer
1 Ea.
Cover Slips
Neubauer
For counting erythrocytes and leukocytes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ Economical Hemacytometer with Cover Slips > Hemacytometer; For use with: Counting erythrocytes and leukocytes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.