Learn More
Hirschmann™ Sodium Heparinized Hematocrit Tubes
Excellent for centrifuging blood in the laboratory. Hirschmann Instruments™ Sodium Heparinized Hematocrit Tubes are manufactured from disposable capillary glass.
Brand:  Hirschmann™ 9100275
 View more versions of this product
Product Code. 10199641
  • 29.45 € / Pack of 1000
Explore more special offers
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Color-coded red
  • Acc. to DIN EN ISO 12772, CE acc. IVD 98/79 EC
Specifications
Specifications
Hematocrit Tube Na-hep
1.07/1.23 mm
75 ±0.5 mm
75 μL
For centrifuging blood samples, Sodium heparinized
DIN EN ISO 12772, CE, IVD 98/79 EC
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products