missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecyltrimethylammonium bromide, 99+%, Thermo Scientific Chemicals
22.51 € - 1663.28 €
Chemical Identifiers
CAS | 57-09-0 |
---|---|
Molecular Formula | C19H42BrN |
Molecular Weight (g/mol) | 364.46 |
MDL Number | MFCD00011772 |
InChI Key | LZZYPRNAOMGNLH-UHFFFAOYSA-M |
PubChem CID | 5974 |
ChEBI | CHEBI:3567 |
SMILES | [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10368600
|
Thermo Scientific Acros
227160100 |
10 g | Glass Bottle |
24.42 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10246520
|
Thermo Scientific Acros
227160250 |
25 g | Plastic Bottle |
22.51 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10645852
|
Thermo Scientific Acros
227161000 |
100 g | Plastic Bottle |
44.52 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10541671
|
Thermo Scientific Acros
227165000 |
500 g | Plastic Bottle |
132.66 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11621789
|
Thermo Scientific Acros
227160025 |
2.5 kg | Plastic Drum |
470.34 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13448488
|
Thermo Scientific Acros
227160101 |
10 kg | Plastic drum |
1663.28 €
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystChemical Identifiers
57-09-0 | |
364.46 | |
LZZYPRNAOMGNLH-UHFFFAOYSA-M | |
CHEBI:3567 |
C19H42BrN | |
MFCD00011772 | |
5974 | |
[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C |
Specifications
White | |
Crystalline Powder | |
>230°C | |
Authentic | |
C19H42BrN | |
MFCD00011772 | |
15,77 | |
Cetrimonium bromide, Cetyltrimethylammonium bromide, CTABr | |
LZZYPRNAOMGNLH-UHFFFAOYSA-M | |
hexadecyltrimethylazanium bromide | |
5974 | |
364.45 | |
Hexadecyltrimethylammonium bromide |
57-09-0 | |
230.0°C | |
244°C | |
99+% | |
CH3(CH2)15N(CH3)3Br | |
04, IV, 819 | |
15, 2024 | |
Solubility in water: 13g/L (20°C). Other solubilities: soluble in methanol (>200g/L at 20 °C), ethanol, insoluble in benzene, diethyl ether, chloroform, sparingly soluble in acetone | |
[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C | |
364.46 | |
CHEBI:3567 | |
≥99% |
Safety and Handling
Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Hexadecyltrimethylammonium bromide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H315-Causes skin irritation.
- H373-May cause damage to organs.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only