missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Hexagonal Antistatic Weighing Boats

Provide accurate pour-outs with minimal sample loss. Fisherbrand™ Hexagonal Antistatic Weighing Boats are flexible polystyrene boats featuring smooth surfaces.

Brand:  Fisherbrand™ HS14251DF

 View more versions of this product

Product Code. 15708197

  • 145.00 € / Pack of 500
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Flexible boats can be bent at points to form pour spouts.
  • Flat bottoms and sloping sides provide accurate transfer.
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • Temperature range -10 to 70°C (14 to 158°F).
  • Can be used with microwave moisture analyzers.
  • Antistatic version only.
  • 1 color, 4 sizes.
  • Pack of 500.
  • All are FDA/food-grade compliant.
  • Disposable.
Specifications

Specifications

Flat
Single-use
Hexagonal
Microwave Moisture Analyzer
89 mm
White
Polystyrene
Antistatic Weighing Boat
127 mm
31 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.