Learn More
Fisherbrand™ Hexagonal Antistatic Weighing Boats
Provide accurate pour-outs with minimal sample loss. Fisherbrand™ Hexagonal Antistatic Weighing Boats are flexible polystyrene boats featuring smooth surfaces.
14.96 € - 145.00 €
Specifications
Bottom Shape Flat
Color White
Disposable Single-use
Material Polystyrene
Shape Hexagonal
View More Specs
Products 4
Product Code Brand Height (Metric) Length (Metric) Width (Metric) Price Quantity & Availability  
Product Code Brand Height (Metric) Length (Metric) Width (Metric) Price Quantity & Availability  
15778187
View Documents
Fisherbrand™
HS14251AF
9 mm 38 mm 25 mm N/A N/A N/A
15788187
View Documents
Fisherbrand™
HS14251BF
19 mm 64 mm 51 mm
63.00 €
Pack of 500
In Stock
to see stock.
Edge
Add to basket
15708197
View Documents
Fisherbrand™
HS14251DF
31 mm 127 mm 89 mm
145.00 €
Pack of 500
Estimated Shipment: 26-05-2025
to see stock.
Add to basket
15798187
View Documents
Fisherbrand™
HS14251CF
22 mm 102 mm 76 mm
113.00 €
Pack of 500
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Flexible boats can be bent at points to form pour spouts.
  • Flat bottoms and sloping sides provide accurate transfer.
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • Temperature range -10 to 70°C (14 to 158°F).
  • Can be used with microwave moisture analyzers.
  • Antistatic version only.
  • 1 color, 4 sizes.
  • Pack of 500.
  • All are FDA/food-grade compliant.
  • Disposable.
Specifications
Specifications
Flat
Single-use
Hexagonal
White
Polystyrene
Microwave Moisture Analyzer
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Hexagonal Antistatic Weighing Boats