missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanes, ≈95% N-Hexane, for HPLC
C6H14, CAS Number-110-54-3, hexanen, hexan, heksan, esani, dipropyl, skellysolve b, hexyl hydride, n-hexane, gettysolve-b, hexanes, 2.5L, 69 deg.C, CHEBI:29021, Colorless, 86.18g/mol, VLKZOEOYAKHREP-UHFFFAOYSA-N, -95 deg.C, 9520, 86.178, Plastic Coated Glass bottle, Liquid, 8058, CCCCCC, 2.97
129.00 € - 307.00 €
Chemical Identifiers
CAS | 110-54-3 |
---|---|
Molecular Formula | C6H14 |
Molecular Weight (g/mol) | 86.18 |
MDL Number | MFCD02179311 |
InChI Key | VLKZOEOYAKHREP-UHFFFAOYSA-N |
Synonym | n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes |
PubChem CID | 8058 |
ChEBI | CHEBI:29021 |
IUPAC Name | hexane |
SMILES | CCCCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10499170
|
Fisher Chemical
H/0406/15 |
1 L | Amber Glass Bottle |
129.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10101910
|
Fisher Chemical
H/0406/PB17 |
2.5 L | Plastic-coated Glass Bottle |
304.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10234150
|
Fisher Chemical
H/0406/21 |
5 L | Aluminum Can |
307.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
C6H14, CAS Number-110-54-3, hexanen, hexan, heksan, esani, dipropyl, skellysolve b, hexyl hydride, n-hexane, gettysolve-b, hexanes, 2.5L, 69 deg.C, CHEBI:29021, Colorless, 86.18g/mol, VLKZOEOYAKHREP-UHFFFAOYSA-N, -95 deg.C, 9520, 86.178, Plastic Coated Glass bottle, Liquid, 8058, CCCCCC, 2.97
Chemical Identifiers
110-54-3 | |
86.18 | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
8058 | |
hexane |
C6H14 | |
MFCD02179311 | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
CHEBI:29021 | |
CCCCCC |
Specifications
-95°C | |
Colorless | |
Liquid | |
MFCD02179311 | |
0.31 mPaS at 20°C | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
hexane | |
8058 | |
86.18g/mol |
110-54-3 | |
69°C | |
C6H14 | |
160mbar at 20°C | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
CCCCCC | |
86.18 | |
CHEBI:29021 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Hexanes, 95% n-Hexane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P273-Avoid release to the environment.