Learn More
IBA Lifesciences Strep-Tactin™ Superflow™ High Capacity Resin
Strep-Tactin™ Superflow™ High Capacity Resin is a 50% suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
Brand:  IBA Lifesciences 2-1208-025
 View more versions of this product
Product Code. 12876456
Description
Description
  • Strep-Tactin™ Superflow™ high capacity is a 6% agarose coupled with the streptavidin variant Strep-Tactin™ and applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
  • In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™ Superflow™ high capacity has a higher density of immobilized Strep-Tactin™, which leads to a 3- to 5-fold higher yield of purified protein.
  • The suspension can be used for packing of gravity flow columns since the resin is optimized for column affinity chromatography, rather than for batch purification.
  • To pack a column with 1 ml resin bed volume, 2 ml 50% suspension is necessary.  
Specifications
Specifications
50 mL
2°C to 8°C
Biotin: > 900 nmol/ mL resin
High Capacity Resin Suspension
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
IBA Lifesciences Strep-Tactin™ Superflow™ High Capacity Resin > Quantity: 50 mL