Learn More
DWK Life Sciences Wheaton™ Crimping Jaw Set for High Performance Crimping Tool
Interchangeable jaws allow users to choose a jaw that best fits their needs.
Brand:  DWK Life Sciences W225761
775.17 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15735148
  • 831.00 € / Each
Explore more special offers
Description
Description
Available as crimper and decappers which fit standard lab vials seals with aluminum or two-part aluminum/steel caps
TRUSTED_SUSTAINABILITY
Specifications
Specifications
High performance crimping tool
13 mm
Crimping
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Crimping Jaw Set for High Performance Crimping Tool > Jaw size: 13mm; Crimping