missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva PhastGel™ Precast IEF Gels
Precast homogeneous gels containing Pharmalyte as carrier ampholyte to generate stable, linear pH gradients with uniform conductivity across the entire pH range and allow high field strengths for high resolution separations.
Brand: Cytiva 17-0544-01
This item is not returnable.
View return policy
Description
- PhastGel IEF gels
- Homogeneous gels in various pH ranges for separations of proteins by isoelectric point

Specifications
125°C | |
PhastGel IEF – 4-6.5 | |
0.45 mm | |
pH 4 to 6.5 | |
Protein | |
Isoelectric Focusing | |
Charecterizing and Analysing Mixtures of Proteins with Smaller PI Differentials | |
82° to 86°C | |
10/Pk. | |
0.017 in. | |
4.7 cm | |
1.9 in. | |
5 cm | |
4.7 cm |
Polyester | |
2 to 6% | |
Pharmalyte | |
IEF Gel | |
pH 4 to 6.5 | |
Containing 2 to 6% Pharmalyte as carrier Ampholyte | |
GelBond polyester film | |
71.08 | |
4 - 8°C | |
50 mm | |
1.9 in. | |
48 mm | |
1.85 in. |
Safety and Handling
Signal Word
Hazard Category
- Danger
Hazard Category
- Germ cell mutagenicity Category 1
- Carcinogenicity Category 1
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva PhastGel™ Precast IEF Gels > IEF 4-6.5
Spot an opportunity for improvement?Share a Content Correction