missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ High Precision Straight Tapered Ultra Fine Point Tweezers/Forceps

Primarily used for microscopy applications. Fisherbrand High Precision Straight Tapered Ultra Fine Point Tweezers/Forceps are made of anti-magnetic stainless steel.

Brand:  Fisherbrand™ 12000122

Product Code. 17210137

  • 28.08 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Primarily used for microscopy applications. Fisherbrand High Precision Straight Tapered Ultra Fine Point Tweezers/Forceps are made of anti-magnetic stainless steel.

  • Straight tapered ultra-fine point
  • Finest points available
  • Made of anti-magnetic stainless steel
Specifications

Specifications

Straight Tapered, Ultra Fine
Stainless Steel
Tweezers/Forceps
0.06 in.
High
9.5 mm
Straight
4.25 in.
11.0 cm
Microscopy
1.5 mm
0.375 in.
Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ High Precision Straight Tapered Ultra Fine Point Tweezers/Forceps > Tweezers/Forceps; Straight tapered ultra fine point

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.