missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen™ Capto™ Q ImpRes and Capto™ SP ImpRes

High-resolution intermediate purification and polishing based on the well-established Capto platform with traditional ligands

Brand:  Cytiva 17-5468-02

464.55 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.



 View more versions of this product

Product Code. 11310002

  • 489.00 € / 100mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Delivers pressure-flow properties with wide resolution
  • High-throughput purifications easy to optimize and scale up
  • High flow agarose based matrix
  • Runs at high flow rates as well as high bed heights thus increasing flexibility in process design
  • Developed for large-scale purification of biopharmaceutical proteins
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
  • Available in a wide range of different bulk pack sizes and pre-packed formats for process development

Capto Q ImpRes: Strong anion (quaternary amine group) exchange BioProcess™ medium
Capto SP ImpRes: Strong cation (sulfonate group) exchange BioProcess medium

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1M NaOH, 8M Urea, 6M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol
39.3 in.
100 mL
200mm
Sulphonate group
4 to 12
High-flow Agarose
High-throughput intermediate purification and polishing steps of a wide range of biomolecules
Capto SP ImpRes medium
1000mm
7.87 in.
220 cm/hr.
<3 bar
36 to 44μm
4° to 30°C
Capto Q Chromatography Media
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Capto SP ImpRes, 100 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.