missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen™ MabSelect™, Xtra™, SuRe™, SuRe LX™ Prepacked Columns

Designed for method optimization and parameter screening for capture of monoclonal antibodies. Cytiva HiScreen™ MabSelect™, Xtra™, SuRe™, SuRe LX™ Prepacked Columns come ready-to-use for convenience.

1037.00 € - 1355.00 €

Specifications

Buffer 8M Urea, 6M Guanidine Hydrochloride, 2% Benzyl Alcohol or 20% Ethanol
Includes Stoppers at the inlet and outlet
Kit Size 1 x 4.7mL
Bed Support Filter
Diameter (English) Inner 0.3 in.
View More Specs

Products 4
Product Code Brand Ligand Product Type Price Quantity & Availability  
Product Code Brand Ligand Product Type Price Quantity & Availability  
10811291
View Documents
Cytiva
28-9269-73
Recombinant Protein A (E.coli) HiScreen MabSelect Prepacked Column
1037.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
10049569
View Documents
Cytiva
28-9269-77
Alkali-stabilized Protein A-derived (E. coli) HiScreen MabSelect SuRe Prepacked Column
1355.00 €
Each
In Stock
to see stock.
Add to basket
 
11603084
View Documents
Cytiva
17-5474-15
Alkali-stabilized Protein A-derived (E. coli) HiScreen MabSelect SuRe LX Prepacked Column
1334.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
10039569
View Documents
Cytiva
28-9269-76
Recombinant Protein A (E.coli) HiScreen MabSelect Xtra Prepacked Column
1037.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Ready-to-use columns for convenient process development
  • Rigid, highly cross-linked agarose matrix
  • Bed height of 10cm is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20cm bed height
  • Small bed volume (4.7mL) gives fast result and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity

MabSelect

  • For capture of MAbs from large sample volumes
  • Prepacked with protein A-derived media
  • Compatible with high flow rates and high pressure when scaling up
MabSelect Xtra
  • Increased binding capacity enables purification of MAbs from high-level expression feedstock
  • Also optimized for Fc-fusion proteins
  • Very low, unspecific binding due to high ligand selectivity and matrix hydrophilicity
  • Prepacked with protein A-derived media
MabSelect SuRe (Superior Resistance) and MabSelect SuRe (Superior Resistance) LX
  • With novel, alkali-tolerant recombinant protein A ligand that provides greater stability and allows use of 0.1 to 0.5M sodium hydroxide for cleaning-in-place (CIP)
  • Enhanced protease resistance resulting in lower ligand leakage
  • High dynamic binding capacity

Specifications

Specifications

8M Urea, 6M Guanidine Hydrochloride, 2% Benzyl Alcohol or 20% Ethanol
1 x 4.7mL
0.3 in.
3.93 in.
Polypropylene
Column
Stoppers at the inlet and outlet
Filter
0.77cm
Epoxy Activation
2° to 8°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiScreen MabSelect

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.