missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ Blue HP Columns
For purifying albumin, enzymes including NAD+ and NADP+, coagulation factors, interferons and related proteins
Brand: Cytiva 17-0412-01
257.86 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
- Prepacked with Blue Sepharose High Performance media
- Cibacron Blue F3G-A, ligand, is covalently attached to the 6% agarose medium
- Stable over pH range 4 to 12 for use and 3 to 13 for cleaning
Specifications
5 x 1mL | |
34 μm | |
1mL | |
0.7cm | |
0.98 in. | |
4mg Cibacron blue F3G-A/mL medium | |
42psi | |
6% Highly Cross-linked Agarose | |
4° to 30°C |
<4 mL/min. | |
Syringe, peristaltic pump or liquid chromato graphy system such as AKTA™ design or FPLC system | |
Blue HP Columns | |
Purifying wide range of biomolecules, including albumin, interferon, a broad range of nucleotide-dependent Enzymes, alpha2-macroglobulin, coagulation facto rs and related Proteins | |
2.5cm | |
Polypropylene | |
4 to 12 | |
Albumin, Interferon, Nucleotide-dependent Enzymes, a2-macroglobulin, Coagulation Factors and Related Proteins |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- hitrap blue HP
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P235-Keep cool.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction