missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Blue HP Columns

For purifying albumin, enzymes including NAD+ and NADP+, coagulation factors, interferons and related proteins

239.00 € - 272.00 €

Specifications

Particle Size 34 μm
For Use With (Equipment) Syringe, peristaltic pump or liquid chromato graphy system such as AKTA™ design or FPLC system
Product Type Blue HP Columns
For Use With (Application) Purifying wide range of biomolecules, including albumin, interferon, a broad range of nucleotide-dependent Enzymes, alpha2-macroglobulin, coagulation facto rs and related Proteins
Height (English) 0.98 in.
View More Specs

Products 2
Product Code Brand Quantity Flow Rate Bed Volume Diameter (Metric) Inner Price Quantity & Availability  
Product Code Brand Quantity Flow Rate Bed Volume Diameter (Metric) Inner Price Quantity & Availability  
10084734
View Documents
Cytiva
17-0412-01
5 x 1mL <4 mL/min. 1mL 0.7cm
272.00 €
Pack of 5
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
11758448
View Documents
Cytiva
17-0413-01
1 x 5mL <20 mL/min. 5mL 1.6cm
239.00 €
Each
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
Description

Description

  • Prepacked with Blue Sepharose High Performance media
  • Cibacron Blue F3G-A, ligand, is covalently attached to the 6% agarose medium
  • Stable over pH range 4 to 12 for use and 3 to 13 for cleaning

Specifications

Specifications

34 μm
Blue HP Columns
0.98 in.
Polypropylene
4 to 12
Albumin, Interferon, Nucleotide-dependent Enzymes, a2-macroglobulin, Coagulation Factors and Related Proteins
Syringe, peristaltic pump or liquid chromato graphy system such as AKTA™ design or FPLC system
Purifying wide range of biomolecules, including albumin, interferon, a broad range of nucleotide-dependent Enzymes, alpha2-macroglobulin, coagulation facto rs and related Proteins
2.5cm
42psi
6% Highly Cross-linked Agarose
4° to 30°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • hitrap blue HP

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Product Certifications