missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ Capto™ MMC ImpRes Column
Prepacked with a weak cation exchange multimodal resin for screening and small-scale purifications
Brand: Cytiva 29401108
This item is not returnable.
View return policy
Description
- The multimodal functionality of Capto MMC ImpRes offers a different selectivity compared to traditional ion exchange columns
- Binds proteins at high or low ionic strength
- Efficiently removes contaminants such as DNA, host cell proteins (HCP), leached protein A, and aggregates
- An excellent choice for purification of MAbs and other biomolecules with high aggregate content.
- Can typically be used in the initial polishing step in MAb purification processes, as well as for polishing of antibody fragments such as domain antibodies (DAbs)
- High yields achieved through the high-resolution beads and selectivity of the ligand
- Enables use of a platform approach to MAb process development
- Displays a broad pH/salt operational window
- Ideal for screening of selectivity, binding, and elution conditions, as well as for small scale purifications

Specifications
High-Resolution Polishing of Monoclonal Antibodies (MAbs) and Other Biomolecules | |
<4 mL/min | |
Multimodal Weak Cation Exchanger | |
60 to 90 mg MAb/mL | |
Pre-packed Column | |
1 mL | |
1 Ea. |
High-Flow Agarose | |
Multimodal Ion Exchange Chromatography | |
Store at 4°C to 30°C in 20% Ethanol, 0.2 M Sodium Acetate | |
5 bar (70 psi) | |
7 mm | |
3 to 12 | |
25 mm |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiTrap Capto MMC ImpRes, 1 x 1 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva HiTrap™ Capto™ MMC ImpRes Column > 1 mL
Spot an opportunity for improvement?Share a Content Correction