missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Heparin HP Columns

Purify DNA-binding proteins, coagulation factors, lipoproteins and protein synthesis factors. Cytiva Life Sciences™ HiTrap™ Heparin HP Columns are prepacked with Heparin Sepharose™ High Performance for high-resolution purification.

Brand:  Cytiva 17-0406-01


 View more versions of this product

Product Code. 10288944

  • 298.00 € / Pack of 5
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stable over pH range 5 to 10
  • Tolerates commonly used aqueous buffers
  • Heparin Sepharose High Performance medium links porcine heparin ligand to 6% agarose by reductive amination

TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 x 1mL
34 μm
1mL
0.7cm
0.98 in.
10mg Heparin/mL medium
42psi
6% Highly Cross-linked Agarose
4° to 30°C
<4 mL/min.
Syringe, peristaltic pump or liquid chromato graphy system such as AKTA™ design or FPLC system
Heparin HP Column
Purification of Antithrombin III, coagulation facto rs and other Plasma Proteins
2.5cm
Polypropylene
5 to 10
DNA-binding Proteins, coagulation facto rs, Lipoproteins and Protein synthesis facto rs
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiTrap Heparin HP 1 ml, 5 x 1 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.