missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Heparin HP Columns

Purify DNA-binding proteins, coagulation factors, lipoproteins and protein synthesis factors. Cytiva Life Sciences™ HiTrap™ Heparin HP Columns are prepacked with Heparin Sepharose™ High Performance for high-resolution purification.

266.00 € - 1165.00 €

Specifications

Particle Size 34 μm
For Use With (Equipment) Syringe, peristaltic pump or liquid chromato graphy system such as AKTA™ design or FPLC system
Product Type Heparin HP Column
For Use With (Application) Purification of Antithrombin III, coagulation facto rs and other Plasma Proteins
Height (English) 0.98 in.
View More Specs

Products 3
Product Code Brand Quantity Flow Rate Diameter (Metric) Inner Price Quantity & Availability  
Product Code Brand Quantity Flow Rate Diameter (Metric) Inner Price Quantity & Availability  
10298944
View Documents
Cytiva
17-0407-01
1 x 5mL <20 mL/min. 1.6cm
266.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
11738448
View Documents
Cytiva
17-0407-03
5 x 5mL <20 mL/min. 1.6cm
1165.00 €
Pack of 5
In Stock
to see stock.
Edge
Add to basket
 
10288944
View Documents
Cytiva
17-0406-01
5 x 1mL <4 mL/min. 0.7cm
298.00 €
Pack of 5
In Stock
to see stock.
Add to basket
 
Description

Description

  • Stable over pH range 5 to 10
  • Tolerates commonly used aqueous buffers
  • Heparin Sepharose High Performance medium links porcine heparin ligand to 6% agarose by reductive amination

Specifications

Specifications

34 μm
Heparin HP Column
0.98 in.
10mg Heparin/mL medium
42psi
6% Highly Cross-linked Agarose
Syringe, peristaltic pump or liquid chromato graphy system such as AKTA™ design or FPLC system
Purification of Antithrombin III, coagulation facto rs and other Plasma Proteins
2.5cm
Polypropylene
5 to 10
4° to 30°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiTrap Heparin HP 5 ml, 1 x 5 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.