missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ rProtein A FF Columns

Use for preparative purification of monoclonal and polyclonal antibodies from cell culture supernatants, serum and ascites. Cytiva Life Sciences™ HiTrap™ rProtein A FF Columns are prepacked with rProtein A Sepharose™ Fast Flow.

Brand:  Cytiva 17-5079-01

Code : GE

Additional Details : Weight : 0.08910kg

 View more versions of this product

Product Code. 10146224

  • 1006.00 € / Pack of 5
Estimated Shipment: 19-07-2024
to see stock.
Add to basket

Description

Description

  • Packed with rProtein A Sepharose™ Fast Flow for purification and fractionation of IgG subclasses and IgG from different sources
  • Exhibits similar Fc region specificity to that of native protein A but with enhanced binding capacity and fewer regulatory concerns due to total absence of mammalian culture in ligand production and purification
  • Average particle size: 90μm
  • Designed for use with a syringe, a peristaltic pump, or a chromatography system
  • Available as 1mL and 5mL columns
Media
  • Ligand: Recombinant protein A (E.coli)
  • Matrix: Highly cross-linked 4% agarose
  • Binding capacity: ∽50mg human IgG/mL
  • pH stability working range: 3 to 10

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.
5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.

Specifications

Specifications

≈50 mg Human IgG/mL
Chromatography System, Manual
4°C to 30°C, 20% Ethanol
<4 mL/min.
For syringe, peristaltic pump or a chromatography system
5 x 1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers