missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ rProtein A FF Columns
Use for preparative purification of monoclonal and polyclonal antibodies from cell culture supernatants, serum and ascites. Cytiva Life Sciences™ HiTrap™ rProtein A FF Columns are prepacked with rProtein A Sepharose™ Fast Flow.
936.00 € - 3782.00 €
Specifications
Binding Capacity | ≈50 mg Human IgG/mL |
---|---|
For Use With (Application) | Chromatography System, Manual |
For Use With (Equipment) | For syringe, peristaltic pump or a chromatography system |
Content And Storage | 4°C to 30°C, 20% Ethanol |
Product Code | Brand | Quantity | Flow Rate | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Flow Rate | Price | Quantity & Availability | ||||
10146224
|
Cytiva
17-5079-01 |
5 x 1 mL | <4 mL/min. |
1006.00 €
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
11708548
|
Cytiva
17-5080-02 |
5 x 5 mL | <20 mL/min. |
3782.00 €
Pack of 5 |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||||
10724547
|
Cytiva
17-5080-01 |
1 x 5 mL | <20 mL/min. |
936.00 €
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
Description
- Packed with rProtein A Sepharose™ Fast Flow for purification and fractionation of IgG subclasses and IgG from different sources
- Exhibits similar Fc region specificity to that of native protein A but with enhanced binding capacity and fewer regulatory concerns due to total absence of mammalian culture in ligand production and purification
- Average particle size: 90μm
- Designed for use with a syringe, a peristaltic pump, or a chromatography system
- Available as 1mL and 5mL columns
- Ligand: Recombinant protein A (E.coli)
- Matrix: Highly cross-linked 4% agarose
- Binding capacity: ∽50mg human IgG/mL
- pH stability working range: 3 to 10
1mL columns
- Bed dimensions: 7 x 25mm
- Flow velocity: <4mL/min.
- Bed dimensions: 16 x 25mm
- Flow velocity: <20mL/min.
Order Info
1mL and 5mL columns are packed with 1mL and 5mL of rProtein A Sepharose Fast Flow, respectively.
Specifications
≈50 mg Human IgG/mL | |
Chromatography System, Manual | |
For syringe, peristaltic pump or a chromatography system | |
4°C to 30°C, 20% Ethanol |