Learn More
HK3 Mouse, anti-Human, Unconjugated, Clone: 3H3C7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67803-1-IG-150UL
This item is not returnable.
View return policy
Description
The HK3 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human HK3. This antibody recognizes human, mouse, rat, pig antigen. The HK3 antibody has been validated for the following applications: IHC, WB,ELISA analysis.
Specifications
HK3 | |
Monoclonal | |
Unconjugated | |
150 μL | |
3101 | |
Liquid |
Western Blot, Immunohistochemistry, ELISA | |
3H3C7 | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |