missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HK3 Mouse, anti-Human, Unconjugated, Clone: 3H3C7, Proteintech
Mouse Monoclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | HK3 |
---|---|
Clone | 3H3C7 |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17211476
|
Proteintech
67803-1-IG-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17201476
|
Proteintech
67803-1-IG-150UL |
150 μL |
399.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The HK3 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human HK3. This antibody recognizes human, mouse, rat, pig antigen. The HK3 antibody has been validated for the following applications: IHC, WB,ELISA analysis.
Specifications
HK3 | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Mouse | |
3101 |
3H3C7 | |
Monoclonal | |
Liquid | |
Human, Mouse, Rat, Pig | |
Primary |