Learn More
Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
Brand:  Hoefer™ SE511-R-1.0
 View more versions of this product
Product Code. 12014807
  • 128.00 € / Each
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Lightweight
  • Virtually unbreakable
  • Come attached to a standard back which produces maximum well depths of 25 mm, except for the 28-well preparative combs which produce well depths of 15 mm
  • Adjustable comb back (available separately, below) converts 25 mm deep combs to 10 or 15 mm depth
Specifications
Specifications
SE 600, SE 400 Series Vertical Electrophoresis System
PTFE
0.03 in.
120μL/ mm
12.1 cm
2.5 cm
Comb
1 Prep, 1 Reference
0.1 cm
4.7 in.
0.98 in.