Learn More
Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
51.60 € - 185.00 €
Specifications
For Use With (Equipment) SE 600, SE 400 Series Vertical Electrophoresis System
Material PTFE
Depth (English) 0.98 in.
Depth (Metric) 2.5 cm
View More Specs
Products 25
Product Code Brand Type Thickness (Metric) Volume (Metric) Well No. of Wells Price Quantity & Availability  
Product Code Brand Type Thickness (Metric) Volume (Metric) Well No. of Wells Price Quantity & Availability  
11579713
View Documents
Hoefer™
SE511-12-1.0
Comb 0.1 cm 7.7μL/ mm 12
113.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
12084867
View Documents
Hoefer™
SE511-DR-1.5
Comb 0.15 cm 171μL/ mm 1 Prep, 2 Reference
128.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
12014707
View Documents
Hoefer™
SE511-DR-.75
Comb 0.07 cm 85μL/ mm 1 Prep, 2 Reference
129.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
12074807
View Documents
Hoefer™
SE511-DR-1.0
Preparative Comb 0.1 cm 112μL/ mm 1 Prep, 2 Reference
128.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
12004877
View Documents
Hoefer™
SE511-20-.75
Standard Comb 0.07 cm 3.1μL/ mm 20
113.00 €
Each
Estimated Shipment: 27-02-2025
to see stock.
Add to basket
10192982
View Documents
Hoefer™
SE511-15-1.5
Standard Comb 0.15 cm 8.6μL/ mm 15
113.00 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
10660036
View Documents
Hoefer™
SE511-20-1,5
Standard Comb 0.15 cm 6.2μL/ mm 20 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12069807
View Documents
Hoefer™
SE511-20-1.5
Standard Comb 0.15 cm 6.2μL/ mm 20
113.00 €
Each
12054647
View Documents
Hoefer™
SE511-15-.75
Standard Comb 0.07 cm 4.3μL/ mm 15
113.00 €
Each
Estimated Shipment: 27-02-2025
to see stock.
Add to basket
12074717
View Documents
Hoefer™
SE511-10-.75
Standard Comb 0.07 cm 6.2μL/ mm 10
113.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
11955348
View Documents
Hoefer™
SE511-12-1.5
Standard Comb 0.15 cm 11.5μL/ mm 12
113.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
11945348
View Documents
Hoefer™
SE511-12-.75
Standard Comb 0.07 cm 5.8μL/ mm 12
113.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
10341135
View Documents
Hoefer™
SE511-10-1.5
Standard Comb 0.15 cm 12.4μL/ mm 10
185.00 €
Each
In Stock
to see stock.
Add to basket
12064757
View Documents
Hoefer™
SE511-10-1.5
Standard Comb 0.15 cm 12.4μL/ mm 10
113.00 €
Each
12089857
View Documents
Hoefer™
SE511-BKA
Adjustable Comb Back
51.60 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
10783847
View Documents
Hoefer™
SE511-BKA
Adjustable Comb Back This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12069817
View Documents
Hoefer™
SE511-R-.75
Comb 0.07 cm 90μL/ mm 1 Prep, 1 Reference
128.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
12014747
View Documents
Hoefer™
SE511-R-1.5
Comb 0.15 cm 183μL/ mm 1 Prep, 1 Reference
128.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
11569723
View Documents
Hoefer™
SE511-28-1.0
Comb 0.1 cm 2.7μL/ mm 28
113.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
12014807
View Documents
Hoefer™
SE511-R-1.0
Comb 0.1 cm 120μL/ mm 1 Prep, 1 Reference
128.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
12074757
View Documents
Hoefer™
SE511-28-.75
Comb 0.07 cm 2.1μL/ mm 28
113.00 €
Each
Estimated Shipment: 28-02-2025
to see stock.
Add to basket
12079807
View Documents
Hoefer™
SE511-28-1.5
Comb 0.15 cm 4.1μL/ mm 28
113.00 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
11539723
View Documents
Hoefer™
SE511-20-1.0
Comb 0.1 cm 4.1μL/ mm 20
113.00 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
12054867
View Documents
Hoefer™
SE511-15-1.0
Comb 0.1 cm 5.7μL/ mm 15
113.00 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
10142892
View Documents
Hoefer™
SE511-10-1.0
Comb 0.1 cm 8.3μL/ mm 10
113.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
Description
Description
  • Lightweight
  • Virtually unbreakable
  • Come attached to a standard back which produces maximum well depths of 25 mm, except for the 28-well preparative combs which produce well depths of 15 mm
  • Adjustable comb back (available separately, below) converts 25 mm deep combs to 10 or 15 mm depth
Specifications
Specifications
SE 600, SE 400 Series Vertical Electrophoresis System
PTFE
0.98 in.
2.5 cm