Learn More
Hoefer™ miniVE Electrophoresis Unit Accessories and Parts
Part and accessories for miniVE Electrophoresis Unit
Brand:  Hoefer™ SE311
 View more versions of this product
Product Code. 11955428
  • 22.15 € / Pack of 5
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accesories include High Voltage Leads (2 pcs), Banana Plug Kit (2 plugs, 2 nuts, 4 washers), Foam Gasket, Clamp Screws (5 pcs), Upper Buffer Chamber Assembly, Clamp Assembly, Sealing Plate Assembly, Sealing Gasket and Lower Buffer Chamber (tank)
Specifications
Specifications
MiniVE Vertical Electrophoresis System
Clamp Screw