missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ miniVE Electrophoresis Unit Accessories and Parts
Part and accessories for miniVE Electrophoresis Unit
22.15 € - 245.00 €
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
11924248
|
Hoefer™
SE314 |
Banana Plug Kit |
42.40 €
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||||
11925428
|
Hoefer™
SE306 |
Lid Assembly |
245.00 €
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
11935428
|
Hoefer™
SE308 |
Lower Buffer Chamber |
80.15 €
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
11975378
|
Hoefer™
SE309 |
Upper Buffer Chamber Assembly |
215.00 €
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
11955428
|
Hoefer™
SE311 |
Clamp Screw |
22.15 €
Pack of 5 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||||
11945428
|
Hoefer™
SE310 |
Clamp Assembly |
67.25 €
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||||
11945358
|
Hoefer™
SE312 |
Sealing Plate Assembly |
39.65 €
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
Description
- Accesories include High Voltage Leads (2 pcs), Banana Plug Kit (2 plugs, 2 nuts, 4 washers), Foam Gasket, Clamp Screws (5 pcs), Upper Buffer Chamber Assembly, Clamp Assembly, Sealing Plate Assembly, Sealing Gasket and Lower Buffer Chamber (tank)
Specifications
MiniVE Vertical Electrophoresis System |