Learn More
Hoefer™ SE 400 Series Sturdier™ Air-Cooled Vertical Electrophoresis Systems
Economical units ideal for applications in which temperature control is not crucial
Brand: Hoefer™ SE410
Includes: Upper Buffer chamber, safety lid with electrodes, lower Buffer chamber and casting stand on leveling base with high voltage Leads, Glass plates, clamp assemblies, cams, slotted gasket for UBC, bubble level, laminated gasket for casting stand, spacer-mate-alignment template, wonder wedge-plate separation tool, gel seal
Requires : Comb and spacers must be ordered separately for Model SE 410 Basic.
Description
- Units run one gel, or two with optional divider plate
- Built-in casting stand seals sandwiches leak-free without tape
- Rugged injection-molded construction for easy assembly and durability in high-use labs
- Plates, clamps, spacers and combs are interchangeable with SE 600 series units
Certifications
EN61010-1, UL3101-1, CSA C22.2 1010.1, CE Marked.
Specifications
EN61010-1, UL3101-1, CSA C22.2 1010.1, CE | |
Up to 80% | |
113°F | |
15°C to 30°C | |
700 mL | |
7.08 in. | |
18 cm | |
9.44 in. | |
5.9 in. | |
500 V | |
60 mA |
18 x 24 cm | |
Upper Buffer chamber, safety lid with electrodes, lower Buffer chamber and casting stand on leveling base with high voltage Leads, Glass plates, clamp assemblies, cams, slotted gasket for UBC, bubble level, laminated gasket for casting stand, spacer-mate-alignment template, wonder wedge-plate separation tool, gel seal | |
45°C | |
23.66 oz. | |
9.44 in. | |
24 cm | |
14.17 in. | |
36 cm | |
15 cm | |
20 w |