Learn More
Sperian™ Honeywell™ 2000 Comfort Series Respirators
With an ergonomic design to deliver outstanding levels of comfort and adjustment. Sperian™ Honeywell™ 2000 Comfort Series Respirators have a cut-out shape to provide excellent compatibility with goggles.
Brand: Sperian™ 1031592
This item is not returnable.
View return policy
Description
- Folds flat packed in individually sealed bags for transportation and storage
- Dolomite tested, demonstrating the respirators' resistance to clogging over time
- With high efficiency exhalation valve for protection
- Double layer face seal provides comfort and protection
Specifications
EN 149 : 2001 A1: 2009 | |
FFP2 | |
Medium | |
Agriculture, Automotive and Part Manufacturer, Building and Construction, Catering, Chemical Industries, Food Industries, Foundry, General Industry, Glass Industries, Green Spaces, Iron and Steel Industry Laboratory, Maintenance Medical and Pharmaceutical | |
2110 FFP2 | |
Flat Fold |
Single-use | |
Respirator Protection | |
Yes | |
Yes | |
25/Pk. |