missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sperian™ Honeywell™ 2000 Comfort Series Respirators
With an ergonomic design to deliver outstanding levels of comfort and adjustment. Sperian™ Honeywell™ 2000 Comfort Series Respirators have a cut-out shape to provide excellent compatibility with goggles.
12.06 € - 41.61 €
Specifications
Certifications/Compliance | EN 149 : 2001 A1: 2009 |
---|---|
Disposable | Single-use |
Product Type | Respirator Protection |
Size | Medium |
With Exhalation Valve | Yes |
Description
- Folds flat packed in individually sealed bags for transportation and storage
- Dolomite tested, demonstrating the respirators' resistance to clogging over time
- With high efficiency exhalation valve for protection
- Double layer face seal provides comfort and protection
Specifications
EN 149 : 2001 A1: 2009 | |
Respirator Protection | |
Yes | |
Flat Fold |
Single-use | |
Medium | |
Yes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title