missing translation for 'onlineSavingsMsg'
Learn More

Wesbart™ Horizational Drawer Rack

Wesbart Horizational Drawer Rack sold complete with 2 drawers.

Brand:  Wesbart™ AH179/2DR

Additional Details : Weight : 3.40000kg

 View more versions of this product

Product Code. 16458299

  • 245.00 € / Each
Estimated Shipment: 18-10-2024
to see stock.
Add to basket

Description

Description

  • Horizational Drawer Rack
  • Sold complete with 2 drawers
Specifications

Specifications

Freezer Rack
2 Drawers
Rectangular
Non-sterile
Freezers
560 mm
560 mm
Aluminum Front Access Rack
Autoclavable
Yes
Storage for Loose Items
415 x 140 x 560 mm
415 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers