Learn More
Thermo Scientific™ Horizon Fog Testing Systems
This system is designed to recreate automotive interior out-gassing in a timely, measurable and repeatable way, utilizing various standards including DIN, ISO and SAE.
Brand:  Thermo Scientific™ 1568005
 View more versions of this product
Product Code. 11917684
  • 10050.00 € / Each
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Thermo Scientific™ Horizon Fog Testing System is designed to recreate automotive interior out-gassing in a timely, measurable and repeatable way, utilizing various standards including DIN, ISO and SAE.
The Horizon Fogging Test System fulfills the DIN, ISO, and SAE standards for fog testing. Available with a choice of controllers, a range of features and a robust pump providing temperature accuracy of ±0.5°C throughout the entire bath ensuring consistent, repeatable results.
  • High capacity heaters provide rapid heating to set point temperature
  • Unique pump nozzle ensures circulation around all six beakers to maintain temperature accuracy
  • Integrated bubble level and four adjusting feet provide easy bath levelling process
  • Sealed frame over bath area prevents heat transfer fluid vapors from corrupting test results
  • Easy-to-view liquid level indicator ensures proper liquid levels are maintained during testing
  • Robust pump design for uniform temperatures
  • Large volume tank wtih 6 flasks enables simultaneous testing, saving valuable time and money
  • Ease of use—drain located on the front of the unit, fill vent on top of the unit, and step-by-step instruction guide
PC-FTS
  • Provides real-time graphs with temperature ramping
  • Displays multiple information
  • Displays data
Ordering Information:
To complete the Fog Tesing System:
  • Choose appropriate cooling system
  • Choose accessory kit for the Gravemetiric or Reflectometric test
  • Select individual accessories
Includes:
Controller, bath, 6 foot power cord
Requires:
Cooling system (Accel 250 LC, SC100-A10 or PC200-A28) and accessories (gravimetric or reflective testing accessory kits)
Services
2 years
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CE/WEE/RoHS
Horizon AC-FTS Fog Testing System
3
Force/suction
Bright LED display
N/A
AC-FTS
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products