Learn More
Product Code | Brand | Model | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | ||||||
11917684
|
Thermo Scientific™
|
AC-FTS |
10050.00 €
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
11927684
|
Thermo Scientific™
|
PC-FTS |
11557.50 €
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
Description
The Thermo Scientific™ Horizon Fog Testing System is designed to recreate automotive interior out-gassing in a timely, measurable and repeatable way, utilizing various standards including DIN, ISO and SAE.
The Horizon Fogging Test System fulfills the DIN, ISO, and SAE standards for fog testing. Available with a choice of controllers, a range of features and a robust pump providing temperature accuracy of ±0.5°C throughout the entire bath ensuring consistent, repeatable results.
- High capacity heaters provide rapid heating to set point temperature
- Unique pump nozzle ensures circulation around all six beakers to maintain temperature accuracy
- Integrated bubble level and four adjusting feet provide easy bath levelling process
- Sealed frame over bath area prevents heat transfer fluid vapors from corrupting test results
- Easy-to-view liquid level indicator ensures proper liquid levels are maintained during testing
- Robust pump design for uniform temperatures
- Large volume tank wtih 6 flasks enables simultaneous testing, saving valuable time and money
- Ease of use—drain located on the front of the unit, fill vent on top of the unit, and step-by-step instruction guide
PC-FTS
- Provides real-time graphs with temperature ramping
- Displays multiple information
- Displays data
Ordering Information:
To complete the Fog Tesing System:
- Choose appropriate cooling system
- Choose accessory kit for the Gravemetiric or Reflectometric test
- Select individual accessories
Includes:
Controller, bath, 6 foot power cord
Requires:
Cooling system (Accel 250 LC, SC100-A10 or PC200-A28) and accessories (gravimetric or reflective testing accessory kits)
Warranty
2 years