missing translation for 'onlineSavingsMsg'
Learn More

Stuart™ Hotplate Accessories

Hotplate Accessories for use with Hotplates/Stirrers that have a 6 x 6 in. top plate.

Brand:  Stuart™ 04805-72

 View more versions of this product

Product Code. 15135934

  • 251.00 € / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • This heating block system with interchangeable inserts can be used with various Stuart Hotplates and Hotplate Stirrers
  • Designed to heat round-bottom flasks
  • Small and medium sized base plates offered; each can accommodate up to three block inserts each
  • A third larger base plate features heat-resistant handles and accepts a single block insert for flasks up to 500 mL
  • Chemical- and solvent-resistant anodized block inserts provide a safe method for synthesis of single or multiple reactions
  • Hotplate accessories offer equal or better heating performance than silicon oil
  • The low-well provides excellent temperature transfer without compromising the view of the reaction or risk of cracking the glassware
  • Each base plate features a hole to house a temperature controller probe
Specifications

Specifications

Hot Plates (6 x 6 in.)
Small
Clamp, Heater Block
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers