missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-21 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 100-42-500UG
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined.Specifications
Q9NSA1 | |
protein with no preservative | |
19.5 | |
500 μg | |
-20°C | |
<1 EU/μg | |
FGF21 | |
Unconjugated | |
MHPIPDSSPL LQFGGQVRQR YLYTDDAQQT EAHLEIREDG TVGGAADQSP ESLLQLKALK PGVIQILGVK TSRFLCQRPD GALYGSLHFD PEACSFRELL LEDGYNVYQS EAHGLPLHLP GNKSPHRDPA PRGPARFLPL PGLPPALPEP PGILAPQPPD VGSSDPLSMV GPSQGRSPSY AS | |
≥ 90% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
26291 | |
Human FGF-21 | |
E. coli | |
RUO | |
FGF; FGF21; FGF-21; FGFL; Fibroblast growth factor; Fibroblast growth factor 21; PRO10196; UNQ3115/PRO10196 | |
Human | |
Recombinant | |
Lyophilized |