missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-21 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
80.50 € - 2980.00 €
Specifications
Accession Number | Q9NSA1 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 26291 |
Molecular Weight (g/mol) | 19.5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17880263
|
Gibco™
100-42-100UG |
100 μg |
598.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17870453
|
Gibco™
100-42-1MG |
1 mg |
2980.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17850763
|
Gibco™
100-42-250UG |
250 μg |
1044.00 €
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17861423
|
Gibco™
100-42-25UG |
25 μg |
195.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17860553
|
Gibco™
100-42-500UG |
500 μg |
1784.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17861323
|
Gibco™
100-42-5UG |
5 μg |
80.50 €
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined.Specifications
Q9NSA1 | |
protein with no preservative | |
19.5 | |
E. coli | |
RUO | |
FGF; FGF21; FGF-21; FGFL; Fibroblast growth factor; Fibroblast growth factor 21; PRO10196; UNQ3115/PRO10196 | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
26291 | |
Human FGF-21 | |
-20°C | |
<1 EU/μg | |
FGF21 | |
Unconjugated | |
MHPIPDSSPL LQFGGQVRQR YLYTDDAQQT EAHLEIREDG TVGGAADQSP ESLLQLKALK PGVIQILGVK TSRFLCQRPD GALYGSLHFD PEACSFRELL LEDGYNVYQS EAHGLPLHLP GNKSPHRDPA PRGPARFLPL PGLPPALPEP PGILAPQPPD VGSSDPLSMV GPSQGRSPSY AS | |
≥ 90% by SDS-PAGE gel and HPLC analyses. |