Learn More
Invitrogen™ Human PAK7 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0368
Description
PAK5 Synthetic Peptide, PEP-0368, from Invitrogen.
PAK7 (KIAA1264) is implicated in the regulation of cytoskeletal dynamics, proliferation, and cell survival signaling. This kinase contains a CDC42/Rac1 interactive binding motif, and has been shown to bind CDC42 in the presence of GTP. This kinase is predominantly expressed in brain. It is capable of promoting neurite outgrowth, and thus may play a role in neurite development. This kinase is associated with microtubule networks and induces microtubule stabilization. The subcellular localization of this kinase is tightly regulated during cell cycle progression. Alternatively spliced transcript variants encoding the same protein have been described.
Specifications
57144 | |
purified | |
Human PAK7 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q9P286 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
PAK5 | |
0.02% sodium azide | |
50 μg | |
PBS |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.